missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution, La2O3 for AAS 0.02mg/L HCl matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
€ 183.00 - € 389.00
Specifications
Concentration | 0.02mg/L |
---|---|
Type | Buffer Solution |
Solvent or Matrix | HCl |
Physical Form | Liquid |
Chemical Name or Material | La2O3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12974545
|
Fisher Chemical
F 001 100 |
100 mL | Bottle |
€ 183.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12984545
|
Fisher Chemical
F 001 500 |
500 mL | Bottle |
€ 389.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
0.02mg/L | |
HCl | |
La2O3 |
Buffer Solution | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Single-element Standard Solution for AAS. Lanthanum (La) 2% in HCl 2%
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to .