Learn More
Invitrogen™ Dual-Light™ System Buffer A
For use with Dual-Light Reporter Gene Assay System. Thermo Scientific Buffer A for Dual-Light Reporter Gene Assay System is supplied in lyophilized form.
Brand:  Invitrogen™ T2176
Product Code. 17194619
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The Dual-Light luminescent reporter gene assay is designed for the rapid and sensitive detection of firefly luciferase and E. coli β-galactosidase in a single extract aliquot for gene expression assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
β-Galactosidase/Luciferase Reporter Gene Assay System
Store the product at -20°C.
Dual-Light™
Solid
Dual-Light™ Reporter Gene Assay System
Room Temperature
200 Assays
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only. Not for use in diagnostic procedures.