Learn More
Eppendorf™ Buckets for Rotor S-4x750
Product Code. 15798369 Shop All Eppendorf Products
Click to view available options
Container Type:
Plate Bucket (Aerosol-tight Capable)
Round Bucket
Quantity:
Set of 2 pcs.
Set of 4 pcs.
Unit Size:
Pack of 2
Pack of 4
Product Code. 15798369
Brand: Eppendorf™ 5895124003
Centrifuge Buckets for Rotor S-4x750
  • Plate bucket S-4x750 2-piece set
  • Round bucket S-4x750 4-piece set
  • Round bucket S-4x750 2-piece set
  • Plate bucket S-4x750 4-piece set
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Rotor S-4x750
Container Type Plate Bucket (Aerosol-tight Capable)
Quantity Set of 4 pcs.