Learn More
BTX™ HT 25-well Electroporation Plates
Click to view available options
Size (Metric) Gap:
2 mm
4 mm
Volume (Metric):
100 μL (Working)
200 μL
Quantity:
1 Ea.
6 Pk.
Unit Size:
Each
Pack of 6
Description
- 25-well disposable electroporation plate
- Standard multi-well formats
- Bio-compatible materials
Specifications
Specifications
| Format | 25-well |
| For Use With (Application) | siRNA Transfection, Library Studies and Stem Cell Project |
| For Use With (Equipment) | ECM 630HT, ECM 830HT, Gemini X2 HT Electroporation System |
| Material | Medical grade polycarbonate, polished aluminium |
| Product Line | BTX Electroporation |
| Product Type | Electroporation consumables |
| Description | 25-Well Disposable Electroporation Plate, 2 mm gap, 100 μl, coated, 1 plate |
| Electroporation Chamber | 25-well Electroporation Plate |
| Includes | 1 x plate with seal |
| Quantity | 1 Ea. |
| Show More |