Learn More
Alpha Packaging™ Brown Laboratory Glassware Pot
Chemically and thermally shockproof. Break resistant.
Brand:  Alpha Packaging™ 280203
 View more versions of this product
Product Code. 10665945
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Brown borosilicate glass
  • Break-resistant
  • Chemically and thermally shockproof;
  • In compliance with EP, USP and JP guidelines
  • Autoclavable
Specifications
Specifications
DIN 45
100 mL
Pot
63 Set
Glass