Learn More
Bromoacetonitrile, 96%, Thermo Scientific Chemicals Product Code.: 11471658
Bromoacetonitrile, 96%, Thermo Scientific Chemicals
Product Code. 11471658
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
Product Code. 11471658
Brand: Thermo Scientific Alfa Aesar A13933.18
Request Bulk or Custom Format
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromoacetonitrile was used in the synthesis of 1-cyanomethyl-1,1-dimethylhydrazinium bromide. It is used as an alkalyting agent for the nitrogen phosphorus detector.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 590-17-0
Molecular Formula C2H2BrN
Molecular Weight (g/mol) 119.95
MDL Number MFCD00001884
InChI Key REXUYBKPWIPONM-UHFFFAOYSA-N
Synonym bromoacetonitrile, acetonitrile, bromo, bromomethyl cyanide, cyanomethyl bromide, ccris 6213, 2-bromoethanenitrile, unii-07k08j16vk, bromacetonitrile, bromoacetonitile, bromo acetonitrileShow MoreShow Less
PubChem CID 11534
IUPAC Name 2-bromoacetonitrile
SMILES BrCC#N
Specifications
Density 1.722
Boiling Point 150°C
Flash Point 81°C (178°F)
Refractive Index 1.48
Quantity 50 g
UN Number UN3276
Beilstein 956569
Solubility Information Insoluble in water.
Formula Weight 119.95
Percent Purity 96%
Chemical Name or Material Bromoacetonitrile
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10239129 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13933.09,A13933.30,A13933.18
Lot Number5014A03Q Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13933.09,A13933.30,A13933.18
Lot Number10144175 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13933.09,A13933.30,A13933.18
Lot Number10174412 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13933.09,A13933.30,A13933.18
Lot NumberUSLF005182 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13933.09,A13933.30,A13933.18
compliance-icons compliance-icons
Product Identifier
  • Bromoacetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Bromoacetonitrile
RUO – Research Use Only
Product Content Correction
Product Title
Bromoacetonitrile, 96%, Thermo Scientific Chemicals >