Learn More
Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals
Product Code. p-4813365 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
Product Code. 11954071
Brand: Thermo Scientific Acros 428721000
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 58.7°C
Chemical Name or Material Bromine
Melting Point -7.2°C
Name Note 1M Solution in trimethyl Phosphate
CAS 7726-95-6, 512-56-1
Quantity 100 mL
Molecular Formula Br
Linear Formula Br2
Flash Point 113°C
Merck Index 15,1401
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
SMILES [Br-]
IUPAC Name molecular bromine
Molecular Weight (g/mol) 79.91
PubChem CID 24408
ChEBI CHEBI:29224
Formula Weight 159.8
Grade ACS Reagent
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.307
Density 1.3070g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromine, 1M solution in trimethyl phosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.