missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ BrdU Staining Kit for Flow Cytometry PE

Product Code. 15591287
Click to view available options
Quantity:
100 tests
Unit Size:
100 tests
This item is not returnable. View return policy
Contains Sodium azide and formaldehyde

Product Code. 15591287

Brand: Invitrogen™ 8812660042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Sodium azide and formaldehyde

Includes

  • BrdU
  • DNase I
  • Anti-BrdU PE Antibody (clone BU20A)
  • BrdU Staining Buffer Concentrate (4X)
  • Fixation/Permeabilization Diluent

Contains the necessary reagents and buffers for identifying and examining proliferating cells of mammalian species by flow cytometric analysis. This kit is optimized to achieve brighter staining compared to traditional protocols.

  • 5μL (0.125μg)/test
  • Cycling cells are incubated with 5-bromo-2′-deoxyuridine (BrdU), a synthetic analog of thymidine which incorporates into newly synthesized genomic DNA during the S-phase of mitosis
  • Following DNA denaturation, the cells are stained for BrdU incorporation along with any other cell surface and/or intracellular targets of interest
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
For Use With (Application) BrdU Staining
For Use With (Equipment) Flow Cytometer
Product Type Staining Kit
Format Kit
Product Line eBioscience
Quantity 100 tests

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.