Learn More
Aalborg™ Brass P-Meter with Optical Sensor Tube
Click to view available options
Length (Metric):
65 mm
150 mm
Unit Size:
Each
Description
- Rib-guided or fluted metering tubes facilitate stable and accurate readings
- Magnifier lens in front shield to enhance reading resolution
- Interchangeability of Flow Tubes and floats
- Ease of installation and exchange of flow tubes
- Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
- Chemical compatibility
- Simple means of panel mounting
- Frame for P-meter with plug
Specifications
Specifications
| For Use With (Equipment) | Flowmeter |
| Type | Support Frame |
| Includes | Brass plug |
| Length (Metric) | 150 mm |
| Width (Metric) | 3.17 mm |
| Warranty | 15Months |