Learn More
Aalborg™ Precision Valve for Brass Flowmeter
Aalborg™ Precision Valve for Brass Flowmeter is designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
€ 120.00
Specifications
For Use With (Equipment) | 150 mm Gas/Liquid Flowmeter |
---|---|
Type | Standard Valve |
Product Code | Brand | Airflow Rate | Waterflow Rate | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Airflow Rate | Waterflow Rate | Price | Quantity & Availability | ||||
11728268
|
Aalborg™
VM1BB |
200 mL/min. | 6mL/min. |
€ 120.00
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
11738268
|
Aalborg™
VM2-BB |
400 mL/min. | 12mL/min. |
€ 120.00
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
||||
11748268
|
Aalborg™
VM3-BB |
1000 mL/min. | 26mL/min. |
€ 120.00
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
11758278
|
Aalborg™
VM5-BB |
6200 mL/min. | 200mL/min. |
€ 120.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
11748278
|
Aalborg™
VM4-BB |
2500 mL/min. | 80mL/min. |
€ 120.00
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
11768278
|
Aalborg™
VM6-BB |
21500 mL/min. | 650mL/min. |
€ 120.00
Each |
Estimated Shipment: 11-03-2025
Log in to see stock. |
||||
Description
- For high precision
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Shells are made of brass, bushings are made of plated brass or 316 stainless steel.
Specifications
150 mm Gas/Liquid Flowmeter | |
Standard Valve |