Learn More
Aalborg™ Brass Flowmeter Kit with Valve
Brand:  Aalborg™ ACRY-010196
 View more versions of this product
Product Code. 11708838
  • € 125.00 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Specifications
Flowmeter Kit
1/8 in. NPT (F)
Brass
±5%
65°C
450 mL/min.
13 LPM
108 mm
25 mm
Full scale
Brass