Learn More
BRAND™ Drying Tube
Product Code. 10439652 Shop All Brand Products
Click to view available options
For Use With (Equipment):
Dispensette digital buret
Unit Size:
Each
Product Code. 10439652
Brand: BRAND™ 707930
Only null left
Add to basket
Add to basket
Fits all models of Dispensette™ Bottle-top Dispensers and Titrette™ Bottle-top Burette
  • Without drying agent
  • Includes valve block sealing ring
TRUSTED_SUSTAINABILITY
Specifications
Product Type Opaque Drying Tube
For Use With (Equipment) Dispensette digital buret