Learn More
Brady™ BradyPrinter i7100 Kit EU with Workstation PWID suite
The BradyPrinter i7100 Industrial Label Printer is an accurate, heavy-duty printer that quickly prints on durable sleeves, printed circuit board labels and asset and component labels.
Brand:  Brady™ 198613
Product Code. 16367595
  • € 2405.00 / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Produce a wide range of high quality identification materials with the BradyPrinter i7100 Kit for EU.
  • Powerful internal processor designed to handle up to 7000 labels per day
  • Intuitive color touch screen interface with distinctive icons to easily access printer menus
  • Optional I/O connection that allows printer to be triggered from programmable controller, foot pedal, sensor or other basic device
  • Center alignment of prints for high printing accuracy
  • Interchangeable platen rollers for optimized printhead lifetime
  • Includes Brady™ Workstation Product & Wire Identification Software Suite
Specifications
Specifications
Color LCD
9.9 in.
11.4 in.
18 in.
Not Autoclavable
Monochrome
Arabic, Bulgarian, Croatian, Czech, Danish, Dutch, English, Finnish, French, German, Greek, Hungarian, Italian, Lithuanian, Norwegian, Polish, Portuguese, Romanian, Russian, Serbian, Slovenian, Spanish, Swedish, Turkish
i7100
600 dpi
Windows™ 10, Windows™ 7, Windows™ 8, Windows™ Servers 2012, 2012 R2, and 2016
22 lb.
Barcode and Inventory Labeling, Electrical Labeling, Laboratory Labeling
110/240 V
Direct Thermal, Thermal Transfer
252 mm
290 mm
460 mm
Non-sterile
i7100 Printer, EU Power Cord, 2 Empty Ribbon Cores, USB Cable, Product CD with Driver, User Manual, Brady™ Workstation Product & Wire ID Suite
256 MB RAM, 50 MB IFFS Flash, SD Card, Slot Up To 512 GB
150 mm/s
Bluetooth™, Ethernet, I/O Connection, Serial, USB
0°C to 40°C
10 kg
2 Year
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers