Learn More
Brady™ BradyJet Laboratory Vinyl Labels
Click to view available options
:
Material: Vinyl
Description
Specifications
Specifications
| Adhesive Type | Acrylic, Permanent |
| Coating | None |
| Color | White |
| Finish | Matte |
| Format | Die-cut |
| Material | Vinyl |
| Printable Sides | Single-side Printable |
| Print Technology | Ink Jet |
| Product Type | Label |
| Sterility | Non-sterile |
| Show More |