Learn More
Brady™ BBP™ Retro Reflective Tape
Product Code. 11796455
Click to view available options
Color:
Orange
Silver
Yellow
Width (English):
1.12 in.
1.125 in.
2.25 in.
4 in.
Unit Size:
Each
Product Code. 11796455
Brand: Brady™ 118002
Only null left
Add to basket
Add to basket
Highly reflective identification that shines back brightly when struck by light
  • Ideal for labeling outdoor utility poles and pedestals, jobsite equipment and roof-top equipment
  • Recommended low light or inclement weather conditions
  • Available in multiple colors
Specifications
Description Retro Reflective Tape for BBP3x Printers
Adhesive Type Acrylic, Pressure-sensitive
Color Yellow
Finish Gloss
Format Tape
Material Polyester
For Use With (Application) For labeling outdoor utility poles and pedestals, jobsite equipment, roof-top equipment
For Use With (Equipment) BBP30, BBP31, BBP33, BBP35, and BBP37 printers
Gloss 100 Gardner Units
Labels per Row 1
Life Span 4 to 6 Years
Quantity 1 Roll
Software Type Brady Workstation, LabelMark, MarkWare
Temperature Range (English) -40 to 158°F
Temperature Range (Metric) -40°C to +70°C
Tensile Strength 10 lb./in.
Thickness (English) 0.0055 in.
Thickness (Metric) 0.14 mm
Length (English) 50 ft.
Length (Metric) 15.24 m
Width (English) 1.12 in.
Width (Metric) 28.58 mm
Alternate Part Number B30C-1125-584-YL