Learn More
Trajan™ BP1 GC Capillary Columns
Click to view available options
Length (Metric):
12 m
12 mm
25 m
30 m
50 m
60 m
60 mm
Diameter (Metric) Inner:
0.15 mm
0.22 mm
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.1 μm
0.25 μm
0.5 μm
1 μm
3 μm
5 μm
0.25 mm
Pakningsstørrelse:
Each
Beskrivelse
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
- Classic cross-linked dimethyl polysiloxane technology
- Low bleed
- Non-polar
- Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Tekniske data
Tekniske data
| Max. Temperature | 300/320°C |
| Functionality/Interaction Type | Van der Waals interaction |
| Diameter (Metric) Inner | 0.53 mm |
| Stationary Phase | 100% Dimethyl Polysiloxane |
| USP Type | G1, G2, G38 |
| For Use With (Application) | Amines Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines |
| Temperature Range | -60°C to +300/320°C |
| Film Thickness | 3 μm |
| Length (Metric) | 30 m |
| Packing Material | Forte™ BP1 |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion