Få mere at vide
Få mere at vide
Corning™ Bottle Top Dispenser (BTD) PTFE Plunger
Shop alle Corning produkter
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
2.5 mL
5 mL
10 mL
25 mL
50 mL
100 mL
Förpackningsstorlek
Each
Beskrivelse
- Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
- Comes with four adapters with different diameters for universal use with multiple bottle sizes
- Telescopic suction tube can be adjusted smoothly to different bottle heights
- Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
- Every dispenser is individually tested and delivered with a certificate of performance
- For dispensing organic and inorganic liquids
Tekniske data
Tekniske data
| Volume (Metric) | 5 mL |
| For Use With (Application) | Dispensing Organic and Inorganic Liquids |
| Includes | Four adapters |