Learn More
Sampling Systems™ Bottle Liquid Sampler
Shop All Sampling Systems Products
Click to view available options
Length (Metric):
1000 mm
1500 mm
Includes:
500 mL Glass Bottle
500 mL Glass Bottle, Bottle Cover
Unit Size:
Each
Description
- 316L Stainless Steel sampler body
- Glass bottle
- Single point
- Sampling for low to medium viscosity liquids
- Ideal for low volume (unit dose) sampling
- The sampler is available with a cover that stops the bottle getting wet
Specifications
Specifications
| Product Type | Liquid Sampler |
| Material | Stainless Steel with Glass Bottle |
| Includes | 500 mL Glass Bottle |
| Capacity (Metric) | 500 mL |
| Length (Metric) | 1000 mm |