Learn More
Cole-Parmer™ Stuart™ Bottle Adapter
Click to view available options
Diameter (Metric) Outer:
33 mm
38 mm
40 mm
45 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Outer | 38 mm |
| For Use With (Equipment) | BT30/50 Digital Burette |
| No. per Pack | 1 Each |
| Type | Buret Adapter |