Learn More
Emma Safety Footwear Boston Safety Shoes
Product Code. 15938391
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
Unit Size:
1 pair
Product Code. 15938391
Brand: Emma Safety Footwear MM40064745
Only null left
Add to basket
Add to basket
The TPU GripForce™ Easy Twist outsole offers maximum grip, is durable and highly recyclable. The integrated lacing prevents pressure points from hooks and eyes and the Hydro-Tec™ lining provides optimal breathability.
  • S1P-Safety shoe with sole plate for dry work
  • Knitted Textile construction
  • Polyurethane coated
  • Hydro-Tec™ Antibacterial
  • Metal-free
  • Chrome-free lining
  • ESD Certified
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black, Gray
Design Boston
Liner Material Hydro-Tec™ Antibacterial
Material Textile
Size 45
Toe Material Composite