Learn More
VITLAB™ Borosilicate Dr. Schilling Buret
Click to view available options
Capacity (Metric):
25 mL
50 mL
Unit Size:
Each
Description
- Patented VITLAB™ symbiotic (DE 10 2005 034 963) impact protection
- Thermally stable plastic coating of the glass buret tube
- Provides additional breaking resistance and acts as a splinter protection
Specifications
Specifications
| Color | Brown |
| Capacity (Metric) | 50 mL |
| Material | Borosilicate Glass |
| Graduations | 0.1 mL |
| Height (Metric) | 900 mm |
| Includes | PE-LD Reservoir Bottle |
| Stopcock Material | PMP, PTFE |
| Tolerance | 0.1 mL |
| Type | Schilling Buret |
| Class | Class B |
| Show More |