Learn More
Kimble Chase™ Borosilicate Glass Plain Chromatography Column
Brand:  Kimble Chase™ 420530-0450
 View more versions of this product
Product Code. 11837483
  • € 413.00 / Each
Estimated Shipment: 24-10-2025
to see stock.
Explore more special offers
Specifications
Ion exchange chromatography
38 mm
Type I
500 mm
With PTFE stopcock plug
1 Each
Plain Chromatography Column
567 mL
Borosilicate Glass
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Kimble Chase™ Borosilicate Glass Plain Chromatography Column > Column Capacity: 567mL