Learn More
Quickfit™ Borosilicate Glass Nitrogen (Kjeldahl) Determination Assembly
Click to view available options
Material:
Pyrex Borosilicate Glass
Unit Size:
Each
Description
Complete assembly for nitrogen determination via the Kjeldahl method. Quickfit™ Borosilicate Glass Nitrogen (Kjeldahl) Determination Assembly is manufactured from heat-resistant and chemically durable borosilicate glass.
- Convenient assembly of Quickfit™ glassware for Kjeldahl digestions and distillations
- Flask can be used alone for digestion stage and then fitted to rest of assembly for the distillation process
- Manufactured from heat resistant and chemically durable borosilicate glass
Specifications
Specifications
| Includes | RA1/22 Adapter receiver, straight typeC1/12/SC Liebig condenserSH7/12 Adapter splash head, vertical distillationMA1/3 Adapter multiple with two parallel necksD3/12 Funnel dropping, open top with Rotaflo™ stopcockFK500/3L Kjeldahl flask 500ml |
| For Use With (Application) | For Nitrogen determination |
| Type | Kjeldahl distillation assembly |
| Material | Pyrex Borosilicate Glass |