Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Gas Distribution Tubes
Brand:  Lenz Laborglasinstrumente™ 07211125
 View more versions of this product
Product Code. 11933564
  • € 41.00 / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Specifications
Borosilicate Glass
1 μm
1 Each
Gas Distribution Tube
250 mm
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Borosilicate Glass Gas Distribution Tubes > Porosity: 1μm