Learn More
Kimble Chase™ Borosilicate Glass Chromatography Column with Fritted Disc and PTFE Stopcock Plug
Click to view available options
Length (Metric):
300 mm
500 mm
600 mm
Diameter (Metric) Inner:
38 mm
50 mm
64 mm
75 mm
missing translation for 'unitSize'
Each
Description
- 40-60 micron porosity glass frit at the lower end to act as a support for column packing material
Specifications
Specifications
| For Use With (Application) | Ion exchange chromatography |
| Quantity | 1 Each |
| Diameter (Metric) Inner | 64 mm |
| Type | Chromatography Column |
| USP Type | Type I |
| Column Capacity | 1608 mL |
| Length (Metric) | 500 mm |
| Material | Borosilicate Glass |
| Includes | With fritted disc and PTFE stopcock plug |