missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boric Acid, Reagent Grade, ≥99.5%, Honeywell™
Reagent Grade,≥99.5%
€ 36.90 - € 371.98
Chemical Identifiers
CAS | 10043-35-3 |
---|---|
Molecular Formula | BH3O3 |
Molecular Weight (g/mol) | 61.83 |
MDL Number | MFCD00011337 |
InChI Key | KGBXLFKZBHKPEV-UHFFFAOYSA-N |
Synonym | orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe |
PubChem CID | 7628 |
ChEBI | CHEBI:33118 |
IUPAC Name | boric acid |
SMILES | OB(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
15687680
|
Honeywell Research Chemicals
B0252-500G |
500 g | Poly Bottle | N/A | ||||||
15667680
|
Honeywell Research Chemicals
B0252-1KG |
1 kg | Poly Bottle | N/A | ||||||
15677680
|
Honeywell Research Chemicals
B0252-2.5KG |
2.5 kg | Poly Bottle |
€ 154.88
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15697680
|
Honeywell Research Chemicals
B0252-5KG |
5 kg | Poly Bottle | N/A | ||||||
15657680
|
Honeywell Research Chemicals
B0252-10KG |
10 kg | Drum with inliner | N/A | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
10043-35-3 | |
61.83 | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
7628 | |
boric acid |
BH3O3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
CHEBI:33118 | |
OB(O)O |
Specifications
3.6 to 4 (at 20°C) | |
10043-35-3 | |
1.510g/cm3 at 20°C | |
2.6mmHg (20°C) | |
H3BO3 | |
MFCD00011337 | |
orthoboric acid, boracic acid, borofax, boron hydroxide, boron trihydroxide, three elephant, basilit b, trihydroxyborone, h3bo3, flea prufe | |
OB(O)O | |
61.83 | |
CHEBI:33118 | |
≥99.5% |
Colorless | |
160°C | |
Odorless | |
BH3O3 | |
Solid | |
NONH for all modes of transport | |
KGBXLFKZBHKPEV-UHFFFAOYSA-N | |
boric acid | |
7628 | |
61.83g/mol | |
Boric acid |
Safety and Handling
P201-P280-P308 + P313
missing translation for 'rtecsNumber' : 233-139-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Boric acid
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Boric Acid, Reagent Grade, ≥99.5%, Honeywell™