Learn More
Berkshire International™ BCR™ Bond Paper
Our BCR™ Bond is made with the purest quality cellulose from specially selected forests and is specially coated to minimize particle and fiber release.
Brand: Berkshire International™ B85A3WH2505
59.95 EUR valid until 2025-05-02
Use promo code "24151" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in 5 colors: Green, Blue, Pink, White, and Yellow
cleanroom, paper, bond
Specifications
White | |
Sheets | |
BCR Bond 850 | |
A3 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
0000850927 | Certificate of Analysis | 11/12/2024 | B85A3WH2505 |
0000850252 | Certificate of Analysis | 08/11/2024 | B85A3WH2505 |
0000850352 | Certificate of Analysis | 04/10/2024 | B85A3WH2505 |
0000850927 | Declaration of Conformity | 11/12/2024 | B85A3WH2505 |
0000850252 | Declaration of Conformity | 08/11/2024 | B85A3WH2505 |
Product Content Correction
Product Title
Berkshire International™ BCR™ Bond Paper > Page Format: A3, White, Bond 850