Learn More
Bohlender™ BOLA™ PTFE Thermometer for Flasks
Designed for use with BOLA flasks and a wide range of reachtion vessels. PTFE thermometers are suited for a variety of applications.
Brand:  Bohlender™ B287-06
Product Code. 10409381
  • € 180.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Description
  • PTFE-jacketed glass thermometer
  • Measuring range 0/+250:1°C
  • Universal chemical resistance
Compliance
FDA
Specifications
Specifications
FDA
600 mm
250°C
0°C to 250°C
7 mm
PTFE
0°C
2,000 to 6,000mL reaction vessel