Learn More
Bohlender™ BOLA™ Polypropylene Tube End Fittings
Different colors for better distinction. Polypropylene Tube fittings with polyamide washers offer very good chemical resistance and are available in a variety of sizes.
Brand:  Bohlender™ F702-32
 View more versions of this product
Product Code. 10639601
  • € 26.00 / Pack of 10
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Colored tube end fittings made of PP
  • Male thread UNF ¼ in. 28 G
  • Washers made of PA are included in delivery
  • Supplied in packs of 10
Compliance
FDA
Specifications
Specifications
FDA
Polypropylene, Polyamide
10/Pk.
UNF 1/4 in.; 28 G
100°C
Tube End Fitting
0°C
1.6 I.D. x 3.2mm O.D. tubing
Green
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers