Learn More
Bohlender™ BOLA™ Magnetic Stirring Bars with Pivot Ring
PTFE-coated magnetic stirring bars with pivot ring.
Brand: Bohlender™ C354-11
This item is not returnable.
View return policy
Description
- PTFE encapsulated magnetic core (Alnico 5)
- Cylindrical shape with pivot ring
- Universal chemical resistance
Specifications
Magnetic Core with Pivot Ring | |
0.8 in. | |
6 mm | |
Cylinder |
0.24 in. | |
PTFE | |
20 mm |