Learn More
Bohlender™ BOLA™ Magnetic Stirring Bars with Pivot Ring
PTFE-coated magnetic stirring bars with pivot ring.
Brand:  Bohlender™ C354-35
 View more versions of this product
Product Code. 17177024
  • € 35.46 / Pack of 3
Estimated Shipment: 31-10-2024
to see stock.
Explore more special offers
Description
  • PTFE encapsulated magnetic core (Alnico 5)
  • Cylindrical shape with pivot ring
  • Universal chemical resistance
Specifications
Specifications
Magnetic Core with Pivot Ring
2.8 in.
9 mm
Cylinder
0.4 in.
PTFE
70 mm