Learn More
Bohlender™ BOLA™ Magnetic Stirring Bars with Pivot Ring
PTFE-coated magnetic stirring bars with pivot ring.
Brand:  Bohlender™ C354-05
 View more versions of this product
Product Code. 17177014
  • € 27.14 / Pack of 10
Estimated Shipment: 31-10-2024
to see stock.
Explore more special offers
Description
  • PTFE encapsulated magnetic core (Alnico 5)
  • Cylindrical shape with pivot ring
  • Universal chemical resistance
Specifications
Specifications
Magnetic Core with Pivot Ring
0.5 in.
5 mm
Cylinder
0.2 in.
PTFE
12 mm