Learn More
Bohlender™ BOLA™ Magnetic Stirring Bars with Pivot Ring
PTFE-coated magnetic stirring bars with pivot ring.
Brand:  Bohlender™ C354-26
 View more versions of this product
Product Code. 17147024
  • € 21.41 / Pack of 3
Estimated Shipment: 31-10-2024
to see stock.
Explore more special offers
Description
  • PTFE encapsulated magnetic core (Alnico 5)
  • Cylindrical shape with pivot ring
  • Universal chemical resistance
Specifications
Specifications
Magnetic Core with Pivot Ring
1.8 in.
8 mm
Cylinder
0.31 in.
PTFE
45 mm