Learn More
Bohlender™ BOLA™ Eco Stopcock Plugs with Bore, Taper 1:10, PTFE
Eco stopcock plugs with nut and grip made of polypropylene, and washer made of PTFE.
Brand:  Bohlender™ J1220-01
Product Code. 17147484
  • € 60.00 / Pack of 3
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Description
  • With bore
  • Smooth surface
  • Grip and nut made of PP
  • Washer included
Specifications
Specifications
1 mm
PTFE
Thread M 7
Taper 1:10
NS 10.0 x 23 mm
Red