Learn More
Thermo Scientific Chemicals Boiling Ston
For analytical work
Brand:  Thermo Scientific Chemicals 386200500
35.40 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10269473
  • € 37.00 / 50g
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Boiling Stone
50 g
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Boiling stones
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466948 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog Number386200500,386202000
Lot NumberA0463527 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog Number386200500,386202000
Lot NumberA0461196 Certificate TypeCertificate of Analysis Date05/06/2024 Catalog Number386200500,386202000
Lot NumberA0455436 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number386200500,386202000
Lot NumberA0447304 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog Number386200500,386202000
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific Chemicals Boiling Ston > 50g; Plastic bottle