Learn More
Grant Instruments™ Test Tube Rack for Unstirred Water Baths
Product Code. p-8014605
Click to view available options
Holds:
105 0.5 mL Tubes
12 30 mm Tubes
18 25 mm Tubes
32 19 mm Tubes
36 16 mm Tubes
55 13 mm Tubes
65 1.5 mL Tubes
84 10 mm Tubes
Unit Size:
Each
Product Code. 11466397
Brand: Grant Instruments™ J2SE
Only null left
Add to basket
Add to basket
Keeps test tubes upright and secure in the bath.
  • Compatible with a wide range of Grant Unstirred Water Baths
  • Holds up to 105 test tubes depending on size
  • Dual slotted trays keep tubes secure and prevent tipping
TRUSTED_SUSTAINABILITY
Specifications
Holds 105 0.5 mL Tubes