missing translation for 'onlineSavingsMsg'
Learn More

Boekel Scientific™ Platform for Microprocessor Controlled Orbital Shaker
GREENER_CHOICE

This is an interchangeable platform for Boekel Orbital Shakers (Cat. No. 15-600-317).

Brand:  Boekel Scientific™ 270292

Product Code. 12663555

  • € 203.00 / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Universal Platform with Adjustable Bars
  • Adjust Bars to Fit Flasks and Beakers of All Sizes
  • interchangeable Locking Design
Specifications

Specifications

2.5 kg
Universal Platform with Adjustable Bars
270 mm
200 mm
Universal
Flasks and Beakers of All Sizes
10.6 in.
7.9 in.
Boekel Microprocessor Controlled Orbital Shaker
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.