Learn More
Buerkle™ Blue Single-Use Tweezers
BIOPHARMAGREENER_CHOICE
Tweezers in a unique shade of blue that does not occur often in foodstuffs, making them easy to find and visually identify.
Brand:  Buerkle™ 5378-1051
 View more versions of this product
Product Code. 16575210
  • € 75.70 / Pack of 100
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Description
  • The use of blue tools thus helps to meet the requirements of the HACCP hazard analysis and assessment in reference to foreign objects
  • This is an important criterion, especially in food processing and production and in the animal feed and pharmaceutical industry
  • The disposable forceps are suitable for precise lab work and are ideal for grasping, lifting and moving small objects, for example fine tissue, small items or solid samples
  • The fluted handles ensure optimum and secure handling, as well as precise work
  • The light plastic forceps for single use allow for fatigue-free work
  • The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
  • The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
  • Individually packaged
  • For single use
  • Clean room manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
  • Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
  • Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
  • Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifications
Specifications
Pointed
5.12 in.
130 mm
Sterile
100
Polystyrene
Forceps
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number244668 Certificate TypeCertificate of Compliance Date10/01/2025 Catalog Number5378-1051
Lot Number244225 Certificate TypeCertificate of Compliance Date19/11/2024 Catalog Number5378-1051
Lot Number241478 Certificate TypeCertificate of Compliance Date24/05/2024 Catalog Number5378-1051
Lot Number231656 Certificate TypeCertificate of Compliance Date03/05/2023 Catalog Number5378-1051
Lot Number224139 Certificate TypeDeclaration of Conformity Date07/12/2022 Catalog Number5378-1051
Special Offers
Special Offers
Product Content Correction
Product Title
Buerkle™ Blue Single-Use Tweezers > Tip Style: Pointed