Learn More
mb2 Chair on Glides - Blue Fabric Upholstered - Asynchronous Mechanism
Click to view available options
:
Height: 660mm
Description
- Chair in fabric blue
- Seat and back in blue fabric upholstered
- Asynchronous mecanism, adjustable seat and backrest
- Pneumatic cylinder Mid
- 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
- Glides
Specifications
Specifications
| Product Type | Contact Seat Chair |
| Color | Blue |
| Adjustable (Height) | Yes |
| Adjustable | Height |
| Back Style | Ergonomic |
| Height (Metric) | 660 mm |
| Holds | 5 Patins |
| Seating Height | 470 to 690 mm |
| Weight (Metric) | 10.22 kg |
| With Armrest | No |
| Show More |