Learn More
Scotch-Brite™ Blue Floor Pads
Product Code. 17128840
Click to view available options
Diameter (Metric):
254 mm
280 mm
305 mm
330 mm
355 mm
380 mm
406 mm
432 mm
460 mm
480 mm
505 mm
530 mm
missing translation for 'unitSize'
Pack of 5
Product Code. 17128840
missing translation for 'mfr': Scotch-Brite™ 7100182697
Only null left
Add to basket
Add to basket
Designed for frequent burnishing with less dust and burning, used to restore the gloss to dull floors. Can be used wet for regular maintenance of unprotected floors. These pads are also recommended for dry stripping soft floors.
  • Heavy-duty scrub pad removes dirt, spills, scuffs, and 1-2 coats of finish
  • Leaves surfaces clean for recoating
  • Designed for frequent burnishing with less dust and burning
  • Washable and reusable pad is economical and helps postpone end-of-life disposal to landfills
  • Versatile pads can be used for heavy-duty spray cleaning and foam scrubbing
  • Available in sizes for rotary and automatic floor machines running at low speed (175-600 rpm)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Stripping Pads
Color Blue
Diameter (Metric) 530 mm
For Use With (Application) Spray Cleaning, and Foam Scrubbing
For Use With (Equipment) Rotary and Automatic Floor Machines
Style Round