Learn More
mb2 Chair on Castors - Blue Fabric Upholstered - Low Lift
Product Code. 11967834
Click to view available options
:
Normal lift
Product Code. 11967834
Brand: mb2 AT1B1D1NE2D6016
Only null left
Add to basket
Add to basket
Work chair with blue fabric - permanent contact - normal lift - castors
  • Chair in fabric blue
  • Seat and back in blue fabric upholstered
  • Backrest mechanism in height, depth and permanent contact adjustable and in every position lockable
  • Pneumatic cylinder low
  • 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
  • Hard floor castors.
Specifications
Product Type Contact Seat Chair
Color Blue
Adjustable (Height) Yes
Adjustable Height
Back Style Ergonomic
Height (Metric) 540 mm
Holds 5 Wheels
Quantity 1 Ea.
Seating Height 440 to 580 mm
Weight (Metric) 9.64 kg
With Armrest No
With Backrest Yes
With Casters Yes
Footrest No
With Lumbar Support No
With Tilt Yes
Material Fabric
Seating Type Desk Height