Learn More
BRAND™ BLAUBRAND™ ETERNA Graduated Pipettes, Type 2, AS, DE-M, AR-GLAS™
All BLAUBRAND™ graduated pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging. On request, they are also available with an individual certificate, USP individual certificate or DAkkS calibration certificate.
€ 61.20 - € 64.80
Description
- DIN EN ISO 835
- Calibrated to deliver (TD, Ex)
- Waiting time 5 seconds
Specifications
Cotton Plug Upper End |