Learn More
BRAND™ BLAUBRAND™ Compact Automatic Burette Spare Burette Tube
Shop All Brand Products
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
Description
- Constructed of Borosilicate Glass 3.3
- Amber glass
Specifications
Specifications
| Color | Amber |
| Capacity (Metric) | 25 mL |
| For Use With (Equipment) | Compact Automatic Burette |
| Material | Borosilicate Glass (3.3) |
| Type | Buret Tube |