Learn More
Brady™ Blank Vinyl Cloth Marker Cards
Brand:  Brady™ 012355
 View more versions of this product
Product Code. 15278304
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Wire Marker Cards - Solid NEMA Colours
Polymer-coated
Matte
Marker Card
900/Pk.
0.055 mm
38 mm
Rubber, Repositionable
Dark Blue
Vinyl Cloth
Wire and cable marking
-40°C to +80°C
6.35 mm