Learn More
mb2 Chair on Castors - Black Fabric Upholstered - Asynchronous Mechanism
Click to view available options
:
Height: 540mm
Description
- Chair in fabric black
- Seat and back in blue fabric upholstered
- Asynchronous mecanism, adjustable seat and backrest
- Pneumatic cylinder low
- 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
- Hard floor castors 50 mm
Specifications
Specifications
| Product Type | Contact Seat Chair |
| Color | Black |
| Adjustable (Height) | Yes |
| Adjustable | Height |
| Back Style | Ergonomic |
| Height (Metric) | 540 mm |
| Holds | 5 Wheels |
| Seating Height | 440 to 560 mm |
| Weight (Metric) | 10.20 kg |
| With Armrest | No |
| Show More |