Learn More
Cytiva BioTrace™ NT Nitrocellulose Transfer Membrane
Click to view available options
Dimensions (LxW):
20 x 20 cm
30 cm x 3 m
7 x 8.5 cm
Unit Size:
1 roll
Pack of 10
Description
- High tensile strength makes it less likely to tear or crack than other brands of nitrocellulose
- High binding capacity for both proteins and nucleic acids
- Ideal for colony/plaque lifts and protein transfers
- Exclusive dot grid pattern does not enhance or inhibit colony growth certified for use in the Membrane Filter (MF) Technique
Specifications
Specifications
| Packaging Type | Sheets |
| Description | Non-sterile |
| Material | BioTrace NT |
| Pore Size | 0.2 μm |
| Dimensions (LxW) | 20 x 20 cm |
| Thickness | 101.6 to 190.5 μm |