Learn More
MedChemExpress Biotin-DADOO
Product Code. 16418778
Encompass
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16418778
Brand: MedChemExpress HYD0980
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
MedChemExpress Biotin-DADOO is a biotinylation reagent, which can be used to synthesize a biotin-estradiol conjugate (i.e., biotin-DADOO-estradiol) to develop a direct, broad range enzyme immunoassay to measure plasma estradiol concentrations.
Specifications
Quantity 10 mg
Product Type Biochemical Assay Reagents
Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
CAS 138529-46-1
Molecular Weight (g/mol) 374.5 g/mol
Solubility DMSO : 100 mg/mL (267.02 mM; Need ultrasonic) ∣ H2O : ≥ 100 mg/mL (267.02 mM)
Synonym Biotinyl-3,6-dioxaoctanediamine ∣ EZ-Link Amine-PEO2-Biotin
Target HBV