Learn More
Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.
Brand: Thermo Scientific™ 75003817
This item is not returnable.
View return policy
Description
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.
- Maximum tube capacity: 4 × 150mL
- Maximum O.D. × L: 50 × 122mm
Specifications
Rotor Tube Adapter | |
25 mL DIN Round-bottom Tube |
4. GP_Multifuge X1_X4 Pro_Megafuge ST1_ST4 Plus_EN_1223.pdf
File size: 3.3 MB
Product Content Correction
Product Title
Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters >