Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Lab Coats and Jackets
- Controlled Environments Lab Coats
- Nitritex™ BioClean-D Disposable Laboratory Coats
-
-
Nitritex Bioclean-DVideo Player is loading.Current Time 0:00/Duration 3:12Loaded: 0%0:00Stream Type LIVERemaining Time -3:121x
- 2x
- 1.75x
- 1.5x
- 1.25x
- 1x, selected
- 0.75x
- 0.5x
- Chapters
- descriptions off, selected
- captions settings, opens captions settings dialog
- captions off, selected
- en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
Nitritex Bioclean-D
Nitritex™ BioClean-D Disposable Laboratory Coats
Product Code. 12942255Product Code. 12942255
Brand: Nitritex™ BDLCXL
Log in to see stock.
Use promo code "24151" to get your promotional price.
Explore more special offers
Description
- Open cuff style
- Three pockets
- CleanTough™ Fabric
- Disposable, packaged in bag
- Spunbond Polypropylene
Specifications
Specifications
Product Type | Controlled Environments Lab Coat |
Size | X-Large |
Material | Double Laminate Microporous Fabric |
Color | White |
Cuff Style | Open |
Disposable | Single-use |
Sterility | Non-sterile |
No. of Pockets | 3 |
Size (Metric) Chest | 1066.8 to 1143 mm |
Certifications/Compliance | ISO 9001:2015, ASTM F739-12 |
Certifications/Compliance (Europe) | (EU)2016/425, EN ISO 13034:2005+A1:2009, EN ISO 14325, EN 1149-5:2008, EN 530, EN 7854, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2 |
Packaging Type | One piece per sealed inner PE bag; one inner bag per sealed outer PE bag; 30 outer bags per lined carton (30 pieces) |
Construction | Spunbond Polypropylene |
Show More Show Less |
Product Suggestions
You may be interested in these alternatives
-
-
-
Product Code. 15248298
-
-
-
-
Product Code. 13569020
-
Product Code. 13589020
Customers who viewed this item also viewed This information does not imply a recommendation or representation of any kind and any action taken upon the information provided is strictly at your own risk. The products shown have not been qualified by Fisher Scientific for any particular purpose, application or use and it is the responsibility of the purchaser to ensure that the performance of the product is suitable for the purchasers specific use or intended application.
-
Catalog No. 15895111
401,00 € / Pack of 50 -
Catalog No. 12901335
278,00 € / Pack of 50 -
Catalog No. 15238298
410,00 € / Pack of 30 -
Catalog No. 15614914
276,00 € / Pack of 100 -
Catalog No. 15278248
303,00 € / Pack of 2000
Videos
-
Nitritex Bioclean-DVideo Player is loading.Current Time 0:00/Duration 3:12Loaded: 0%0:00Stream Type LIVERemaining Time -3:121x
- 2x
- 1.75x
- 1.5x
- 1.25x
- 1x, selected
- 0.75x
- 0.5x
- Chapters
- descriptions off, selected
- captions settings, opens captions settings dialog
- captions off, selected
- en (Main), selected
This is a modal window.
Beginning of dialog window. Escape will cancel and close the window.
End of dialog window.
This is a modal window. This modal can be closed by pressing the Escape key or activating the close button.
Nitritex Bioclean-D