Learn More
Angelantoni Industrie™ Bioban Microbiological Safety Cabinets
Routinely used in laboratory of microbiology, virology, hematology and cell cultures.
Brand: Angelantoni Industrie™ ST-003034000
This item is not returnable.
View return policy
Description
- Working chamber of stainless steel AISI 304L 2B finish
- Side walls made of tempered safety glass
- Fluorescent lamp 36w
- Double inlet centrifugal type fan with external rotor motor and degree of protection IP 55
- Recirculated air filtration and expulsion carried out with HEPA H14 filters
Compliance
EN12469
Specifications
EN 12469 | |
99.995% | |
Microbiology, Virology, Hematology and Cell Cultures |
BIOBAN 48 | |
HEPA H14 Filters |